Anti-ABHD5 Goat Polyclonal Antibody

Supplier: ProSci
45-186
10115-616EA 701.94 USD
10115-616
Anti-ABHD5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 38 kDa band observed in lysates of cell lines A431 and NIH3T3 (calculated MW of 39.1 kDa according to NP_057090.2). Recommended concentration: 1-3 ?g/mL.

ABHD5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABHD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Cow
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR