Anti-ABCE1 Goat Polyclonal Antibody

Supplier: ProSci
45-183
10115-614EA 701.94 USD
10115-614
Anti-ABCE1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 70 kDa band observed in lysates of cell lines A431 (calculated MW of 67.6 kDa according to NP_002931.1). Recommended concentration: 0.3-1 ?g/mL.

ABCE1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR