Anti-EIF4ENIF1 Goat Polyclonal Antibody

Supplier: ProSci
45-177
10115-604EA 701.94 USD
10115-604
Anti-EIF4ENIF1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1/32,000. Western Blot: Approximately 140-150 kDa band observed in 293 lysates (predicted size of approximately 108 kDa according to NP_062817.1, however our observation agrees with that of Dostie et al (see below). Recommended for use at 0.25-0.5 ?g/mL. Immunohistochemistry: In paraffin embedded human pancreas shows strong staining of cytoplasm in acinar cells. Recommended concentration 5-10 ?g/mL.

4E-T antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EIF4ENIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR