Anti-XRCC5 Goat Polyclonal Antibody

Supplier: ProSci
45-173
10115-598EA 701.94 USD
10115-598
Anti-XRCC5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 80-85 kDa band observed in lysates of HeLa (calculated MW of 82.7 kDa according to NP_066964.1). Recommended concentration: 0.1-0.3 ?g/mL.

XRCC5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: XRCC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR