Anti-UBE2I Goat Polyclonal Antibody

Supplier: ProSci
45-164
10115-582EA 701.94 USD
10115-582
Anti-UBE2I Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 17 kDa band observed in human kidney, human lung and human testis lysates (calculated MW of 18 kDa according to NP_003336). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human kidney, shows nuclear staining of some epithelial cells of glomeruli and renal tubules. Recommended concentration, 1 ?g/mL.

UBE2I antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: UBE2I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR