Anti-STAT3 Goat Polyclonal Antibody

Supplier: ProSci

45-154
10112-274EA 560.36 USD
10112-274
Anti-STAT3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 90 kDa band observed in lysates of NIH3T3 (calculated MW of 88.0 kDa according to NP_644805.1 and of 88.1 according to NP_003141.2). Recommended concentration: 1-3 ?g/mL. Immunohistochemistry: Stains mouse thymic medulla and not the cortex. Recommended overnight staining at 10 ?g/mL.

STAT3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: STAT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR