Anti-SH3GLB1 Goat Polyclonal Antibody

Supplier: ProSci
45-143
10115-550EA 701.94 USD
10115-550
Anti-SH3GLB1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 38 kDa band observed in lysates of human T cell line Jurkat and in human lung and placenta (calculated MW of 40.8 kDa according to NP_057093.1). Recommended concentration: 0.3-1.0 ?g/mL. A minor band of unknown identity was also consistently observed at 50 kDa in Jurkat and placenta. This band was successfully blocked by incubation with the immunizing peptide.

BIF-1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SH3GLB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR