Anti-RBP1 Goat Polyclonal Antibody

Supplier: ProSci
45-136
10115-536EA 701.94 USD
10115-536
Anti-RBP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 16 kDa band observed in Murine NIH 3T3 lysates (calculated MW of 15.9 kDa according to NP_002890.1). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human pancreas shows vesicular staining in the acinar cytoplasm but oriented towards the interlobular space. Recommended concentration, 5-10 ?g/mL.

RBP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR