Anti-RANBP1 Goat Polyclonal Antibody

Supplier: ProSci
45-134
10115-532EA 701.94 USD
10115-532
Anti-RANBP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 28-30 kDa band observed in Hela and HepG2 lysates (predicted MW of 23 kDa according to NP_002873). Recommended for use at 0.1-1 ?g/mL.

RANBP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RANBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR