Anti-RAC2 Goat Polyclonal Antibody

Supplier: ProSci
45-131
10115-526EA 701.94 USD
10115-526
Anti-RAC2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 20 kDa band observed in lysates of human tonsil lysate (calculated MW of 21.4 kDa according to NP_002863.1). Recommended concentration: 0.1-1 ?g/mL. Immunohistochemistry: In paraffin embedded human tonsil, stains most cells in the mantle zone and interfollicular area. In the germinal centres the majority of cells are negative. Recommended concentration 0.3 ?g/mL. IHC data were obtained with a previous batch.

RAC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR