Anti-COX1 Goat Polyclonal Antibody

Supplier: ProSci
45-124
10115-516EA 701.94 USD
10115-516
Anti-COX1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 60 kDa and 70 kDa bands observed in HeLa lysate (calculated MW of 64.5 kDa according to NP_542158.1 and 68.7 kDa according to NP_000953.2) Recommended for use at 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human heart shows patchy staining of myocardial fibres in longitudinal section. Recommended concentration, 3-5 ?g/mL.

COX-1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PTGS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR