Anti-POU2AF1 Goat Polyclonal Antibody

Supplier: ProSci
45-116
10115-502EA 701.94 USD
10115-502
Anti-POU2AF1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 30 kDa band seen in mouse spleen lysate (Predicted MW of 28 kDa according to NP_006226). Recommended for use at 1-3 ?g/mL.

POU2AF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POU2AF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR