Anti-PDIA3 Goat Polyclonal Antibody

Supplier: ProSci
45-108
10115-486EA 701.94 USD
10115-486
Anti-PDIA3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 55 kDa band observed in human hepatoblastoma HepG2 lysates (calculated MW of 56.8 kDa according to NP_005304.3). Recommended concentration: 1-3 ?g/mL.

PDIA3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDIA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR