Anti-PAX5 Goat Polyclonal Antibody

Supplier: ProSci
45-105
10115-480EA 701.94 USD
10115-480
Anti-PAX5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 50 kDa band seen in U937 lysate (predicted MW 45 kDa ). Recommended for use at 2-4 ?g/mL.

PAX5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PAX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR