Anti-NPM Goat Polyclonal Antibody

Supplier: ProSci

45-099
10115-468EA 560.36 USD
10115-468
Anti-NPM Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 37 kDa band observed in human HeLa lysates (calculated MW of 32.6 kDa according to NP_002511.1 and 29.5 kDa according to NP_954654.1). Recommended concentration: 0.1-0.3 ?g/mL.

NucleopH osmin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NPM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR