Anti-MSN Goat Polyclonal Antibody

Supplier: ProSci
45-093
10115-456EA 701.94 USD
10115-456
Anti-MSN Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 70 kDa band observed in human spleen lysates (calculated MW of 67.8 kDa according to NP_002435.1). Recommended concentration: 1-3 ?g/mL.

Moesin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MSN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR