Anti-MMP7 Goat Polyclonal Antibody

Supplier: ProSci
45-090
10115-450EA 701.94 USD
10115-450
Anti-MMP7 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 28 kDa band observed in human breast, ovary and placenta lysates (calculated MW of 29.7 kDa according to NP_002414.1). Recommended concentration: 0.03-0.1 ?g/mL.

MMP-7 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MMP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR