Anti-MELK Goat Polyclonal Antibody

Supplier: ProSci
45-089
10115-448EA 701.94 USD
10115-448
Anti-MELK Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: experiments gave bands at approximately 75 kDa and 150 kDa in human testis lysates and in cell lines HeLa and Jurkat after 0.1 ?g/mL antibody staining. The150 kDa band is explained by glycosylation of this member of the AMPK family. This protein has a calculated MW of 74.6 kDa according to NP_055606.1. Recommended concentration: 0.1-0.3 ?g/mL.

MELK antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MELK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR