Anti-MAP4K1 Goat Polyclonal Antibody

Supplier: ProSci
45-086
10115-442EA 701.94 USD
10115-442
Anti-MAP4K1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1/32,000. Western Blot: Approximately 95-100 kDa band observed in Jurkat (predicted MW of 96 kDa according to NP_009112). Recommended for use at 0.5-2 ?g/mL.

MAP4K1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAP4K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR