Anti-LIME Goat Polyclonal Antibody

Supplier: ProSci
45-085
10115-440EA 701.76 USD
10115-440
Anti-LIME Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution >1:128,000. Western Blot: Approximately 32 kDa band observed in mouse heart lysates (predicted MW of 31 kDa according to NP_076173.1). Recommended for use at 0.5-2 ?g/mL.

LIME1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Lime1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR