Anti-LDLRAP1 Goat Polyclonal Antibody

Supplier: ProSci
45-082
10115-434EA 701.94 USD
10115-434
Anti-LDLRAP1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 40-45 kDa band observed in human liver lysate (predicted MW of 35 kDa according to NP_056442.1). Recommended for use at 1-3 ?g/mL.

LDLRAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LDLRAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR