Anti-JUP Goat Polyclonal Antibody

Supplier: ProSci
45-076
10115-422EA 701.94 USD
10115-422
Anti-JUP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 85 kDa band observed in A431 lysate (predicted MW of 86 kDa according to NP_002221, NP_068831). Recommended for use at 0.2-1 ?g/mL. Enzyme immunoassay: Customer finds it successful in quantitative ELISA assays (EIA) but the antibody is very sensitive to freeze/thaw cycles in this assay.

JUP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: JUP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR