Anti-HPGD Goat Polyclonal Antibody

Supplier: ProSci
45-070
10115-410EA 701.94 USD
10115-410
Anti-HPGD Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Experiments gave bands at approximately 26 kDa and 50 kDa in human duodenum lysates after 0.1 ?g/mL antibody staining. These bands correspond to earlier findings in literature with different antibodies (PMID: 11889207). This protein has a calculated MW of 29.0 kDa according to NP_000851. Recommended concentration: 0.1-1 ?g/mL. Immunohistochemistry: In paraffin embedded human colon shows strong cytoplasm staining of enterocytes. Recommended concentration, 2-4 ?g/mL.

HPGD antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HPGD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR