Anti-GRB2 Goat Polyclonal Antibody

Supplier: ProSci
45-066
10115-402EA 701.94 USD
10115-402
Anti-GRB2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 30 kDa band seen in Jurkat lysate. Recommended for use at 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human tonsil, stains mantle zone and to a lessser extent germinal centre. Recommended concentration, 0.1-0.3 ?g/mL.

GRB2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GRB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR