Anti-GFAP Goat Polyclonal Antibody

Supplier: ProSci
45-063
10115-396EA 701.94 USD
10115-396
Anti-GFAP Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 48 kDa band observed in mouse brain lysates (calculated MW of 49.8 kDa according to mouse NP_034407.2 and human NP_002046.1). Recommended concentration: 1-3 ?g/mL.

GFAP antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR