Anti-FLOT1 Goat Polyclonal Antibody

Supplier: ProSci
45-051
10115-382EA 701.94 USD
10115-382
Anti-FLOT1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 48 kDa band observed in lysates of cell line H460 (calculated MW of 47.4 kDa according to NP_005794.1). In transfected HEK293 transiently expressing FLOT1 a band of approximately 48 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended for use at 0.3-1 ?g/mL.

Flotillin 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FLOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR