Anti-CHAT Goat Polyclonal Antibody

Supplier: ProSci
45-037
10112-252EA 701.94 USD
10112-252
Anti-CHAT Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 75-80 kDa band observed in mouse and rat brain lysates (calculated MW of 72.3 kDa according to NP_034021.1). Recommended concentration: 0.2-0.5 ?g/mL.

ChAT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Chat
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR