Anti-CBX3 Goat Polyclonal Antibody

Supplier: ProSci
45-029
10115-346EA 701.76 USD
10115-346
Anti-CBX3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 20 kDa band observed in lysates of A431, HeLa and HepG2. (calculated MW of 20.8 kDa according to NP_009207.2 and NP_057671.2). Recommended concentration: 0.3-1.0 ?g/mL. Immunohistochemistry: In paraffin embedded human placenta shows strong nuclear staining in trophoblasts. Recommended concentration, 2-3 ?g/mL.

CBX3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CBX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR