Anti-BAIAP2 Goat Polyclonal Antibody

Supplier: ProSci

45-017
10112-250EA 560.36 USD
10112-250
Anti-BAIAP2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 55 kDa band observed in human brain (cerebellum and frontal cortex) lysates (calculated MW of 60.9 kDa according to NP_006331.1). Recommended concentration: 1-3 ?g/mL. an additional band of unknown identity was also consistently observed at 150-170 kDa . This band was successfully blocked by incubation with the immunizing peptide.

BAIAP2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BAIAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR