Anti-AURKA Goat Polyclonal Antibody

Supplier: ProSci
45-014
10115-322EA 701.94 USD
10115-322
Anti-AURKA Goat Polyclonal Antibody
Antibodies
  • ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 45 kDa band seen in Jurkat and HeLa cell lysates. Recommended for use at 0.5-2 μg/mL.

Aurora Kinase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Aurora kinase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AURKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR