Anti-ATF4 Goat Polyclonal Antibody

Supplier: ProSci
45-013
10115-320EA 701.94 USD
10115-320
Anti-ATF4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1000. Western Blot: Approximately 40 kDa band seen in Raji cell lysate. Recommended for use at 1-3 ?g/mL.

ATF4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR