Anti-ARPC2 Goat Polyclonal Antibody

Supplier: ProSci
45-011
10115-316EA 701.76 USD
10115-316
Anti-ARPC2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 33-34 kDa band observed in Hela and human skeletal muscle lysates (predicted MW of 34 kDa according to NP_005722). Recommended for use at 0.1-0.5 ?g/mL. Immunohistochemistry: In paraffin embedded human spleen shows cytoplasm staining in some splenocytes in and around a T-cell area. Recommended concentration, 3-5 ?g/mL.

ARPC2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARPC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR