Anti-RIMS2 Rabbit Polyclonal Antibody

Supplier: ProSci

4445
10061-902EA 441.03 USD
10061-902
Anti-RIMS2 Rabbit Polyclonal Antibody
Antibodies
RIM2 Antibody: Rab3-interacting molecules (RIMs) are synaptic proteins necessary for neuronal transmission and plasticity. RIM1 and RIM2 proteins are expressed in overlapping but distinct patterns throughout the brain. While the ablation of either gene was not lethal in mice, the deletion of both resulted in postnatal mortality. This lethality is due to a defect in neurotransmitter release; synapses without RIM proteins can still release neurotransmitters but are unable to do so in response to normal Ca2+ triggers. Like RIM1, RIM2 is thought to be an effector protein for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner.

RIM2 antibody can be used for detection of RIM2 by immunohistochemistry at 5 ?g/mL.

RIM2 Antibody is affinity chromatography purified via peptide column.

RIM2 Antibody is supplied in PBS containing 0.02% sodium azide.

RIM2 antibody can be stored at 4°C for three months and -20°C, stable for up to one year.

Type: Primary
Antigen: RIMS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR