Anti-GPX1 Goat Polyclonal Antibody

Supplier: ProSci
43-088
10112-224EA 701.94 USD
10112-224
Anti-GPX1 Goat Polyclonal Antibody
Antibodies
The starting concentration recommended for Western Blot is 0.1-0.3?g/mL.

Gpx1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Gpx1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR