Anti-FBL Goat Polyclonal Antibody

Supplier: ProSci
43-079
10113-480EA 701.94 USD
10113-480
Anti-FBL Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128000. The starting concentration recommended for Western Blot is 0.1-0.3?g/mL.

FBL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR