Anti-B2M Goat Polyclonal Antibody

Supplier: ProSci

43-039
10113-460EA 560.36 USD
10113-460
Anti-B2M Goat Polyclonal Antibody
Antibodies
The starting concentration recommended for Western Blot is 0.3-1?g/mL.

B2M antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR