Anti-S100 A8 Goat Polyclonal Antibody

Supplier: ProSci
43-038
10115-216EA 701.94 USD
10115-216
Anti-S100 A8 Goat Polyclonal Antibody
Antibodies
The starting concentration recommended for Western Blot is 0.03-0.1?g/mL.

S100A8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR