Anti-TRIM6 Goat Polyclonal Antibody

Supplier: ProSci
42-989
10115-168EA 552.02 USD
10115-168
Anti-TRIM6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 8000. Western Blot: Preliminary experiments gave an approx 70-75 kDa band in Human Kidney and Liver lysates after 0.2 ?g/mL antibody staining.

TRIM6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRIM6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR