Anti-CCNDBP1 Goat Polyclonal Antibody

Supplier: ProSci
42-984
10115-166EA 701.94 USD
10115-166
Anti-CCNDBP1 Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 40 kDa band observed in Human Placenta lysates (calculated MW of 40.3 kDa according to NP_036274.3). Recommended concentration: 0.3-1 ?g/mL.

CCNDBP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCNDBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR