Anti-CXCR2 Goat Polyclonal Antibody

Supplier: ProSci
42-981
10115-160EA 701.94 USD
10115-160
Anti-CXCR2 Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 40 kDa band observed in Mouse Colon lysates (calculated MW of 40.4 kDa according to NP_034039.1). Recommended concentration: 0.3-1 ?g/mL.

Cxcr2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cxcr2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR