Anti-DBH Goat Polyclonal Antibody

Supplier: ProSci
42-975
10115-154EA 701.94 USD
10115-154
Anti-DBH Goat Polyclonal Antibody
Antibodies
Western Blot: Approx 70 kDa band observed in Jurkat lysate (calculated MW of 69.1 kDa according to NP_000778.3). Recommended concentration: 1-3 ?g/mL.

DBH antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DBH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR