Anti-ABCC9 Goat Polyclonal Antibody

Supplier: ProSci

42-972
10115-152EA 455.3 USD
10115-152
Anti-ABCC9 Goat Polyclonal Antibody
Antibodies
Western Blot: Preliminary experiments gave an approx 90 kDa band in Human Testis lysates and 100 kDa in Rat Testis lysates after 0.1 ?g/mL antibody staining.

ABCC9 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR