Anti-Clu Goat Polyclonal Antibody

Supplier: ProSci
42-962
10113-396EA 701.94 USD
10113-396
Anti-Clu Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 8000. Western Blot: Approximately 37 kDa band observed in mouse and rat testis lysates (calculated MW of 51.7 kDa according to NP_038520.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Miyata et al, Circulation. 2001 Sep 18;104 (12) :1407-12.; PMID: 11560857). Recommended concentration: 0.1-0.3 ?g/mL.

Clu antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Clu
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR