Anti-MCM5 Goat Polyclonal Antibody

42-953
10113-386EA 389.35 USD
10113-386
Anti-MCM5 Goat Polyclonal Antibody
ELISA: Antibody detection limit dilution 32000

mcm5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MCM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR