Anti-KCNC3 Goat Polyclonal Antibody

Supplier: ProSci
42-949
10113-380EA 701.94 USD
10113-380
Anti-KCNC3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 64000. Western Blot: Approximately 80 kDa band observed in mouse fetal brain lysates (calculated MW of 81.9 kDa according to NP_032448.2). Recommended concentration: 0.2-0.6 ?g/mL.
Some minor background is detected and is blocked by the immunizing peptide. We call for caution when used for other assays than Western Blot.

Kcnc3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Kcnc3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR