Anti-GPR83 Goat Polyclonal Antibody

Supplier: ProSci

42-936
10113-364EA 560.36 USD
10113-364
Anti-GPR83 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 2000. Western Blot: Approximately 50 kDa band observed in human brain (cerebellum, cerebral cortex and amygdala) lysates (calculated MW of 48.3 kDa according to NP_057624.3). Recommended concentration: 1-3 ?g/mL.

GPR83 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR