Anti-KCNC3 Goat Polyclonal Antibody

Supplier: ProSci
42-935
10113-362EA 701.94 USD
10113-362
Anti-KCNC3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 16000. Western Blot: Approximately 80 kDa band observed in mouse and rat brain lysates (calculated MW of 81.9 kDa according to mouse NP_032448.2). Recommended concentration: 0.3-1 ?g/mL.

KCNC3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KCNC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR