Anti-C16orf57 Goat Polyclonal Antibody

Supplier: ProSci
42-933
10115-120EA 701.94 USD
10115-120
Anti-C16orf57 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 16000. Western Blot: Approximately 35+28 kDa bands observed in human, mouse and rat skin lysates (calculated MW of 30.3 kDa according to NP_078874.2 and 28.1 kDa according to NP_001182231.1). Recommended concentration: 0.3-1 ?g/mL.

C16orf57 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: C16orf57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR