Anti-PCDH17 Goat Polyclonal Antibody

Supplier: ProSci
42-932
10115-118EA 701.94 USD
10115-118
Anti-PCDH17 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 8000. Western Blot: Approximately 150 kDa band observed in mouse fetal brain lysates (calculated MW of 126 kDa according to human NP_001035519.1 and mouse NP_001013775.2). Recommended concentration: 0.1-0.3 ?g/mL.

PCDH17 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PCDH17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR