Anti-NGFR Goat Polyclonal Antibody

Supplier: ProSci

42-927
10115-108EA 560.36 USD
10115-108
Anti-NGFR Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 4000. Western Blot: Approximately 75 kDa band observed in rat heart lysates (calculated MW of 45.2 kDa according to human NP_002498.1 and 45.4 kDa according to rat NP_036742.2. Recommended concentration: 1-3 ?g/mL.

NGFR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NGFR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR